Please login to your online account to display your discounted pricing

CD25 Mouse anti-Human, Clone: GLLZDMY, eBioscience™

Mouse Monoclonal Antibody

$101.50 - $265.35

Specifications

Antigen CD25
Clone GLLZDMY
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122638

 
life technologies
LS14025680
25μg Each for $101.50

501122639

 
life technologies
LS14025682
100μg Each for $265.35
Description & Specifications

Specifications

Antigen CD25
Clone GLLZDMY
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Human
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This GLLZDMY monoclonal antibody reacts with human CD25, which is also known as the low-affinity interleukin (IL)-2 receptor alpha. CD25 is expressed on activated T cells (including a subset of regulatory T cells), B cells, and macrophages. This receptor subunit associates with CD122, the IL-2 receptor beta chain, and CD132, the common gamma chain, to form the high-affinity IL-2 receptor. IL-2 receptor signaling has been linked to cell proliferation and survival in lymphocytes. CD25 is a marker for Hodgkin’s disease, and has been associated with such autoimmune diseases such as Type I diabetes.