missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD25 Mouse anti-Canine, eFluor 660, Clone: P4A10, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $328.68

Specifications

Antigen CD25
Clone P4A10
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra, IL2Ra
Species Reactivity Canine
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016431 Life Technologies
LS50025041
25 Tests Each for $130.68
5016432 Life Technologies
LS50025042
100 Tests Each for $328.68
Specifications

Description

This P4A10 monoclonal antibody reacts with canine CD25, the 55kDa interleukin-2 receptor alpha (IL-2Rα) chain. As in humans and mice, CD25 in dogs is expressed by early T and B lineage progenitors as well as by activated mature T and B lymphocytes. CD25 associates with the IL-2R beta (CD122) and gamma (CD132) chains to form the high-affinity IL-2 receptor. On its own, CD25 binds IL-2 with low affinity. Along with CD4 and Foxp3, CD25 is a major marker used to define regulatory T cells.

The P4A10 antibody has been reported to recognize the same epitope as the anti-human CD25 monoclonal antibody ACT-1, but with greater affinity.
Description & Specifications

Specifications

CD25
Murine
Canine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
P4A10
Interleukin-2 Receptor alpha, IL-2Ra, IL2Ra
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test