Please login to your online account to display your discounted pricing

CD247 Mouse anti-Human, PE, Clone: 6B10.2, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $462.55

Specifications

Antigen CD247
Clone 6B10.2
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010739

 
life technologies
LS12247980
25μg Each for $187.05

5010740

 
life technologies
LS12247982
100μg Each for $462.55
Description & Specifications

Specifications

Antigen CD247
Clone 6B10.2
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This 6B10.2 monoclonal antibody reacts with human CD247, which is also known as the invariant zeta chain. The CD3 complex is composed of three monomeric chains (γ, δ, and ε) and the invariant zeta chain, which exists as a homodimer or as a heterodimer with the η subunit. Together with the T cell antigen receptor (TCR), the CD3 γ, δ, ε, and zeta chains comprise the TCR/CD3 complex. CD3 zeta is expressed on immature and mature thymocytes, as well as T lymphocytes and NK cells. Signal transduction during T cell activation is mediated by tyrosine phosphorylation of cytoplasmic immunoreceptor tyrosine-based activation motifs (ITAMs) present on the zeta chain.

The 6B10.2 monoclonal antibody recognizes a transmembrane epitope of the CD3 zeta chain. Therefore, flow cytometric analysis with this antibody requires intracellular staining. In addition, this antibody has been reported to recognize the zeta chain as a homodimer but not as a monomer.