Please login to your online account to display your discounted pricing

CD244 Mouse anti-Human, FITC, Clone: eBioDM244, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $288.55

Specifications

Antigen CD244
Clone eBioDM244
Host Species Mouse
Gene Alias 2B4, p38, SLAMF4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122143

 
life technologies
LS11583741
25 tests Each for $143.55

50245244

 
life technologies
LS11583742
100 tests Each for $288.55
Description & Specifications

Specifications

Antigen CD244
Clone eBioDM244
Host Species Mouse
Gene Alias 2B4, p38, SLAMF4
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The eBioDM244 monoclonal antibody reacts with human CD244 (2B4, p38). In human, CD244 is a 38kDa protein expressed on NK cells, a subset of CD8+ T cells, gamma delta T cells, monocytes, basophils and eosinophils. Binding of the CD244 ligand, CD48, results in NK cell activation, unlike mouse CD244, which is an inhibitory receptor.