missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD244 Mouse anti-Human, Clone: eBioPP35 (PP35), eBioscience™

Mouse Monoclonal Antibody

$85.55 - $158.05

Specifications

Antigen CD244
Clone eBioPP35 (PP35)
Host Species Murine
Gene Alias 2B4, p38, SLAMF4
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013000 Life Technologies
LS14244980
25μg Each for $85.55
5013001 Life Technologies
LS14244982
100μg Each for $158.05
Specifications

Description

The eBioPP35 monoclonal antibody reacts with human CD244, also known as 2B4, p38, NK-cell activation-inducing ligand, signaling lymphocyte activation molecule family member 4, and natural killer cell receptor 2B4. Human CD244 is a 38 kD protein expressed on NK cells, a subset of CD8+ T cells, γδ T cells, monocytes, basophils, mast cells, and eosinophils. Binding of CD244 to its ligand, CD48, results in NK cell activation. Costaining experiments with eBioC1.7 suggest that both antibodies recognize different epitopes.
Description & Specifications

Specifications

CD244
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioPP35 (PP35)
2B4, p38, SLAMF4
Flow Cytometry,Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL