Please login to your online account to display your discounted pricing

CD244 Mouse anti-Human, Biotin, Clone: eBioC1.7 (C1.7), eBioscience™

Mouse Monoclonal Antibody

$100.05 - $172.55

Specifications

Antigen CD244
Clone eBioC1.7 (C1.7)
Host Species Murine
Gene Alias 2B4, p38, SLAMF4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011944

 
life technologies
LS13583881
50μg Each for $100.05

5011945

 
life technologies
LS13583882
100μg Each for $172.55
Description & Specifications

Specifications

Antigen CD244
Clone eBioC1.7 (C1.7)
Host Species Murine
Gene Alias 2B4, p38, SLAMF4
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The eBioC1.7 monoclonal antibody reacts with human CD244 (2B4, p38). In human, CD244 is a 38kDa protein expressed on NK cells, a subset of CD8+ T cells, γδ T cells, monocytes, basophils and eosinophils. Binding of the CD244 ligand, CD48, results in NK cell activation, unlike mouse CD244, which is an inhibitory receptor. For CD244 expressed on NK cells, binding of CD48 results in enhanced NK cell cytotoxicity and secretion of IFN-γ. Recently, it was demonstrated that binding of the C1.7 monoclonal antibody and CD48 involve the same residue in the V domain of human CD244, which explains the ability of C1.7 binding to induce activation of NK cells. Binding of C1.7 to CD244 leads to tyrosine phosphorylation and recruitment of the adaptor molecule SAP (SLAM-associated protein). Patients with X-linked lymphoproliferative disorder (XLPD) have a mutation in SAP which renders it unable to bind to phosphorylated CD244.