Please login to your online account to display your discounted pricing

CD24 Rat anti-Mouse, PerCP-eFluor 710, Clone: M1/69, eBioscience™

Rat Monoclonal Antibody

$158.05 - $477.05

Specifications

Antigen CD24
Clone M1/69
Host Species Rat
Gene Alias Heat Stable Antigen, HSA
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015937

 
life technologies
LS46024280
25μg Each for $158.05

5015938

 
life technologies
LS46024282
100μg Each for $477.05
Description & Specifications

Specifications

Antigen CD24
Clone M1/69
Host Species Rat
Gene Alias Heat Stable Antigen, HSA
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The M1/69 monoclonal antibody reacts with the mouse CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. CD24 is a variably glycosylated molecule resulting in heterogeneity of molecular mass of this antigen on cells of different lineages and antibodies to CD24 exhibit subtle differences in staining level on lymphocyte populations. The expression of CD24 has been used to resolve stages of B lymphopoiesis in mouse bone marrow. It has been reported that P-selectin (CD62P) binds to CD24.