missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD24 Rat anti-Mouse, PE-Cy5, Clone: M1/69, eBioscience™

Rat Monoclonal Antibody

$196.68 - $341.88

Specifications

Antigen CD24
Clone M1/69
Host Species Rat
Gene Alias Heat Stable Antigen, HSA
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014029 Life Technologies
LS15024281
50μg Each for $196.68
5014030 Life Technologies
LS15024282
100μg Each for $341.88
Specifications

Description

The M1/69 monoclonal antibody reacts with the mouse CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. CD24 is a variably glycosylated molecule resulting in heterogeneity of molecular mass of this antigen on cells of different lineages and antibodies to CD24 exhibit subtle differences in staining level on lymphocyte populations. The expression of CD24 has been used to resolve stages of B lymphopoiesis in mouse bone marrow. It has been reported that P-selectin (CD62P) binds to CD24.
Description & Specifications

Specifications

CD24
Rat
Flow Cytometry
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
M1/69
Heat Stable Antigen, HSA
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer