Please login to your online account to display your discounted pricing

CD24 Mouse anti-Human, PE-Cy7, Clone: eBioSN3 (SN3 A5-2H10), eBioscience™

Mouse Monoclonal Antibody

$191.40 - $471.25

Specifications

Antigen CD24
Clone eBioSN3 (SN3 A5-2H10)
Host Species Mouse
Gene Alias Heat Stable Antigen, HAS
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123271

 
life technologies
LS25024741
25 tests Each for $191.40

501123272

 
life technologies
LS25024742
100 tests Each for $471.25
Description & Specifications

Specifications

Antigen CD24
Clone eBioSN3 (SN3 A5-2H10)
Host Species Mouse
Gene Alias Heat Stable Antigen, HAS
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The eBioSN3 (SN3 A5-2H10) monoclonal antibody reacts with the human CD24 molecule, also known as Heat Stable Antigen (HAS). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. It has been reported that P-selectin (CD62P) binds to CD24.