missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD24 Mouse anti-Human, PE, Clone: eBioSN3 (SN3 A5-2H10), eBioscience™

Mouse Monoclonal Antibody

$143.88 - $303.60

Specifications

Antigen CD24
Clone eBioSN3 (SN3 A5-2H10)
Host Species Murine
Gene Alias Heat Stable Antigen, HSA
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010283 Life Technologies
LS12024741
25 Tests Each for $143.88
501129573 Life Technologies
LS12024742
100 Tests Each for $303.60
Specifications

Description

The SN3 monoclonal antibody reacts with the human CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. It has been reported that P-selectin (CD62P) binds to CD24.
Description & Specifications

Specifications

CD24
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
eBioSN3 (SN3 A5-2H10)
Heat Stable Antigen, HSA
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer