missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD24 Mouse anti-Human, Clone: eBioSN3 (SN3 A5-2H10), eBioscience™

Mouse Monoclonal Antibody

$77.88 - $143.88

Specifications

Antigen CD24
Clone eBioSN3 (SN3 A5-2H10)
Host Species Murine
Gene Alias Heat Stable Antigen, HSA
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012484 Life Technologies
LS14024780
25μg Each for $77.88
5012485 Life Technologies
LS14024782
100μg Each for $143.88
Specifications

Description

The SN3 monoclonal antibody reacts with the human CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. It has been reported that P-selectin (CD62P) binds to CD24.
Description & Specifications

Specifications

CD24
Murine
Flow Cytometry
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
eBioSN3 (SN3 A5-2H10)
Heat Stable Antigen, HSA
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer