Please login to your online account to display your discounted pricing

CD24 Mouse anti-Human, Biotin, Clone: eBioSN3 (SN3 A5-2H10), eBioscience™

Mouse Monoclonal Antibody

$100.05 - $216.05

Specifications

Antigen CD24
Clone eBioSN3 (SN3 A5-2H10)
Host Species Murine
Gene Alias Heat Stable Antigen, HSA
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122505

 
life technologies
LS13024782
100μg Each for $216.05

5011481

 
life technologies
LS13024780
25μg Each for $100.05
Description & Specifications

Specifications

Antigen CD24
Clone eBioSN3 (SN3 A5-2H10)
Host Species Murine
Gene Alias Heat Stable Antigen, HSA
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The SN3 monoclonal antibody reacts with the human CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. It has been reported that P-selectin (CD62P) binds to CD24.