missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD24 Mouse anti-Human, APC, Clone: eBioSN3, eBioscience™

Mouse Monoclonal Antibody

$174.24 - $443.52

Specifications

Antigen CD25
Clone PC61.5
Host Species Rat
Gene Alias Interleukin-2 Receptor alpha
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014921 Life Technologies
LS17024741
25 Tests Each for $174.24
5014922 Life Technologies
LS17024742
100 Tests Each for $443.52
501129526 Life Technologies
LS17025182
100μg Each for $368.28
Specifications

Description

The SN3 monoclonal antibody reacts with the human CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. It has been reported that P-selectin (CD62P) binds to CD24.
Description & Specifications

Specifications

CD25
Rat
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
PC61.5
Interleukin-2 Receptor alpha
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer