missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ CD24 (GPI-Linked Surface Mucin) Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures.

$304.31 - $1,417.63

Specifications

Antigen CD24 (GPI-Linked Surface Mucin) Ab-2
Clone SN3b
Applications Flow Cytometry,Immunohistochemistry (Paraffin),Flow Cytometry,Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS1279P Thermo Scientific™
MS1279P
200μg/mL 1mL Each for $1,364.71
MS1279P0 Thermo Scientific™
MS1279P0
200μg/mL 100μL Each for $304.31
MS1279P1 Thermo Scientific™
MS1279P1
200μg/mL 500μL Each for $854.94
MS1279P1ABX Thermo Scientific™
MS1279P1ABX
1mg/mL 100μL Each for $964.88
MS1279PABX Thermo Scientific™
MS1279PABX
1mg/mL 200μL Each for $1,417.63
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures with Thermo Scientific™ CD24 (GPI-Linked Surface Mucin) Ab-2, Mouse Monoclonal Antibody.

CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. It is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation.

Host Species: Mouse

Clone: SN3b

Isotype: IgM/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line

Molecular Weight: 30-70 kDa

Positive Control: Breast CA, Ovarian CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Note:

USA: IVD; Int'l: IVD

Description & Specifications

Specifications

CD24 (GPI-Linked Surface Mucin) Ab-2
Flow Cytometry,Immunohistochemistry (Paraffin),Flow Cytometry,Immunohistochemistry (Paraffin)
Unlabeled
Concentrated
RUO
Primary
IgM kappa
SN3b
Monoclonal
Human
Murine
B-Cell
Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line