Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD24 (GPI-Linked Surface Mucin) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, and flow cytometry experiments with Thermo Scientific™ CD24 (GPI-Linked Surface Mucin) Ab-1.

 

$244.00 - $850.22

Specifications

Antigen CD24 (GPI-Linked Surface Mucin) Ab-1
Clone SN3
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1278P

 
thermo scientific™ lab vision™
MS1278P
200μg/mL 1mL Each for $750.98

MS1278P0

 
thermo scientific™ lab vision™
MS1278P0
200μg/mL 100µL Each for $244.00

MS1278P1

 
thermo scientific™ lab vision™
MS1278P1
200μg/mL 500µL Each for $589.20

MS1278P1ABX

 
thermo scientific™ lab vision™
MS1278P1ABX
1mg/mL 100µL Each for $677.73

MS1278PABX

 
thermo scientific™ lab vision™
MS1278PABX
1mg/mL 200µL Each for $850.22
Description & Specifications

Specifications

Antigen CD24 (GPI-Linked Surface Mucin) Ab-1
Clone SN3
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline B-Cell
Type Primary
Immunogen Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line
Isotype IgG2a kappa

Ensure accurate, reproducible results in immunohistochemistry, western blotting, and flow cytometry experiments with Thermo Scientific™ CD24 (GPI-Linked Surface Mucin) Ab-1.

CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. It is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation.

Host Species: Mouse

Clone: SN3

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line

Molecular Weight: 30-70 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO