Please login to your online account to display your discounted pricing

CD235a (Glycophorin A) Mouse anti-Human, PerCP-eFluor 710, Clone: HIR2 (also GA-R2), eBioscience™

Mouse Monoclonal Antibody

$249.40 - $530.70

Specifications

Antigen CD235a (Glycophorin A)
Clone HIR2 (also GA-R2)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016119

 
life technologies
LS46998741
25 Tests Each for $249.40

5016120

 
life technologies
LS46998742
100 Tests Each for $530.70
Description & Specifications

Specifications

Antigen CD235a (Glycophorin A)
Clone HIR2 (also GA-R2)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.002μg)/test

The HIR2 monoclonal antibody reacts with human glycophorin A, a sialoglycoprotein expressed by erythroid precursors and mature circulating red cells.