Please login to your online account to display your discounted pricing

CD235a (Glycophorin A) Mouse anti-Human, PE, Clone: HIR2 (also GA-R2), eBioscience™

Mouse Monoclonal Antibody

$129.05 - $317.55

Specifications

Antigen CD235a (Glycophorin A)
Clone HIR2 (also GA-R2)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011348

 
life technologies
LS12998782
100μg Each for $317.55

501129468

 
life technologies
LS12998780
25μg Each for $129.05
Description & Specifications

Specifications

Antigen CD235a (Glycophorin A)
Clone HIR2 (also GA-R2)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The HIR2 monoclonal antibody reacts with human glycophorin A, sialoglycoproteins expressed by erythroid precursors and mature circulating red cells.