missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD235a (Glycophorin A) Mouse anti-Human, Clone: HIR2 (also GA-R2), eBioscience™

Mouse Monoclonal Antibody

$64.68 - $130.68

Specifications

Antigen CD235a (Glycophorin A)
Clone HIR2 (also GA-R2)
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013970 Life Technologies
LS14998780
25μg Each for $64.68
5013971 Life Technologies
LS14998782
100μg Each for $130.68
Specifications

Description

The HIR2 monoclonal antibody reacts with human glycophorin A, a sialoglycoprotein expressed by erythroid precursors and mature circulating red cells.
Description & Specifications

Specifications

CD235a (Glycophorin A)
Murine
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HIR2 (also GA-R2)
Flow Cytometry
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL