missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD235a (Glycophorin A) Mouse anti-Human, Biotin, Clone: HIR2 (also GA-R2), eBioscience™

Mouse Monoclonal Antibody

$105.60 - $242.88

Specifications

Antigen CD235a (Glycophorin A)
Clone HIR2 (also GA-R2)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012311 Life Technologies
LS13998780
25μg Each for $105.60
501128741 Life Technologies
LS13998782
100μg Each for $242.88
Specifications

Description

The HIR2 monoclonal antibody reacts with human glycophorin A, a sialoglycoprotein expressed by erythroid precursors and mature circulating red cells.
Description & Specifications

Specifications

CD235a (Glycophorin A)
Murine
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
HIR2 (also GA-R2)
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer