Please login to your online account to display your discounted pricing

CD23 Rat anti-Mouse, FITC, Clone: B3B4, eBioscience™

Rat Monoclonal Antibody

$129.05 - $404.55

Specifications

Antigen CD23
Clone B3B4
Host Species Rat
Gene Alias Low Affinity IgE Receptor, FceRII
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509490

 
life technologies
LS11023281
50μg Each for $129.05

509491

 
life technologies
LS11023282
100μg Each for $230.55

509492

 
life technologies
LS11023285
500μg Each for $404.55
Description & Specifications

Specifications

Antigen CD23
Clone B3B4
Host Species Rat
Gene Alias Low Affinity IgE Receptor, FceRII
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The B3B4 monoclonal antibody reacts with mouse CD23, a 45kDa type II transmembrane glycoprotein. CD23 is expressed on resting conventional B cells, and its expression is modulated upon B-cell activation. B-1 cell lineage (CD5+ B cells) does not express CD23. Soluble forms of the antigen have been reported to be biologically active. CD23 is a low affinity receptor for IgE and is thought to play a role in the regulation of IgE response and B-cell activation. CD21 is thought to bind to CD23.