missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD23 Rat anti-Mouse, eFluor 660, Clone: B3B4, eBioscience™

Rat Monoclonal Antibody

$117.48 - $289.08

Specifications

Antigen CD23
Clone B3B4
Host Species Rat
Gene Alias Low Affinity IgE Receptor, FceRII
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124260 Life Technologies
LS50023280
25μg Each for $117.48
501124261 Life Technologies
LS50023282
100μg Each for $289.08
Specifications

Description

The B3B4 monoclonal antibody reacts with mouse CD23, a 45kDa type II transmembrane glycoprotein. CD23 is expressed on resting conventional B cells, and its expression is modulated upon B-cell activation. B-1 cell lineage (CD5+ B cells) does not express CD23. Soluble forms of the antigen have been reported to be biologically active. CD23 is a low affinity receptor for IgE and is thought to play a role in the regulation of IgE response and B-cell activation. CD21 is thought to bind to CD23.
Description & Specifications

Specifications

CD23
Rat
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
B3B4
Low Affinity IgE Receptor, FceRII
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer