missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD23 Mouse anti-Human, PE-Cy7, Clone: EBVCS2, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $394.68

Specifications

Antigen CD23
Clone EBVCS2
Host Species Murine
Gene Alias Low Affinity IgE Receptor, FceRII, FcER2, IGEBF
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015481 Life Technologies
LS25023841
25 Tests Each for $157.08
5015482 Life Technologies
LS25023842
100 Tests Each for $394.68
Specifications

Description

The EBVCS2 monoclonal antibody reacts with human CD23, a 45kDa type II transmembrane glycoprotein. CD23 is expressed on mature B cells, mantle zone B cells, follicular dendritic cells and at low levels on T, NK, langerhans cells and platelets. Expression of CD23 is upregulated upon B cell activation, and soluble forms of the antigen have been reported to be biologically active. CD23 is a low affinity receptor for IgE and is thought to play a role in the regulation of IgE response and B cell activation. CD21 and the alpha subunit of CD11b and CD11c bind to CD23.
Description & Specifications

Specifications

CD23
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
EBVCS2
Low Affinity IgE Receptor, FceRII, FcER2, IGEBF
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test