missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD23 Mouse anti-Human, eFluor 450, Clone: EBVCS2, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $328.68

Specifications

Antigen CD23
Clone EBVCS2
Host Species Murine
Gene Alias Low Affinity IgE Receptor, FceRII, FcER2, IGEBF
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124096 Life Technologies
LS48023842
100 Tests Each for $328.68
501129318 Life Technologies
LS48023841
25 Tests Each for $130.68
Specifications

Description

The EBVCS2 monoclonal antibody reacts with human CD23, a 45kDa type II transmembrane glycoprotein. CD23 is expressed on mature B cells, mantle zone B cells, follicular dendritic cells and at low levels on T, NK, langerhans cells and platelets. Expression of CD23 is upregulated upon B cell activation, and soluble forms of the antigen have been reported to be biologically active. CD23 is a low affinity receptor for IgE and is thought to play a role in the regulation of IgE response and B cell activation. CD21 and the alpha subunit of CD11b and CD11c bind to CD23.
Description & Specifications

Specifications

CD23
Murine
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
EBVCS2
Low Affinity IgE Receptor, FceRII, FcER2, IGEBF
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer