Please login to your online account to display your discounted pricing

CD23 Mouse anti-Human, Clone: EBVCS2, eBioscience™

Mouse Monoclonal Antibody

$85.55 - $158.05

Specifications

Antigen CD23
Clone EBVCS2
Host Species Mouse
Gene Alias Low Affinity IgE Receptor, FceRII, FcER2, IGEBF
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012476

 
life technologies
LS14023880
25μg Each for $85.55

5012477

 
life technologies
LS14023882
100μg Each for $158.05
Description & Specifications

Specifications

Antigen CD23
Clone EBVCS2
Host Species Mouse
Gene Alias Low Affinity IgE Receptor, FceRII, FcER2, IGEBF
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The EBVCS2 monoclonal antibody reacts with human CD23, a 45kDa type II transmembrane glycoprotein. CD23 is expressed on mature B cells, mantle zone B cells, follicular dendritic cells and at low levels on T, NK, langerhans cells and platelets. Expression of CD23 is upregulated upon B cell activation, and soluble forms of the antigen have been reported to be biologically active. CD23 is a low affinity receptor for IgE and is thought to play a role in the regulation of IgE response and B cell activation. CD21 and the alpha subunit of CD11b and CD11c bind to CD23.