missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD226 (DNAM-1) Rat anti-Mouse, Clone: 10E5, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$216.05 - $549.55

Specifications

Antigen CD226 (DNAM-1)
Clone 1.00E+06
Host Species Rat
Gene Alias DNAM1, PTA1, Tlisa
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014505 Life Technologies
LS16226181
50μg Each for $216.05
5014506 Life Technologies
LS16226185
500μg Each for $549.55
Specifications

Description

This 10E5 monoclonal antibody reacts with mouse CD226, which is also known as DNAX accessory molecule-1 (DNAM-1). CD226 is an adhesion molecule expressed primarily on T cells, including thymocytes. The highest levels of CD226 are found on naïve CD8 T cells. Moreover, expression of CD226 is regulated differentially in CD4 T cells. Under certain culture conditions, CD226 can be upregulated in Th1 and downregulated in Th2 and Th0 cells. This cell surface receptor can also be detected on natural killer cells, some B cell subsets, monocytes, and platelets. Interaction with its ligands, the poliovirus receptor (CD155) and CD112, results in adhesion and cell signaling events that promote innate and adaptive immunity, including differentiation, survival, and cytotoxicity.

The 10E5 monoclonal antibody has been reported to inhibit antigen-specific T cell proliferation and IFN-gamma production in vivo.
Description & Specifications

Specifications

CD226 (DNAM-1)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, no sodium azide
1.00E+06
DNAM1, PTA1, Tlisa
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL