missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD223 (Lag-3) Mouse anti-Human, eFluor 450, Clone: 3DS223H, eBioscience™

Mouse Monoclonal Antibody

$200.64 - $483.12

Specifications

Antigen CD223 (Lag-3)
Clone 3DS223H
Host Species Murine
Gene Alias Lymphocyte Activation Gene-3
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124149 Life Technologies
LS48223942
100 Tests Each for $483.12
501129290 Life Technologies
LS48223941
25 Tests Each for $200.64
Specifications

Description

This 3DS223H monoclonal antibody recognizes human CD223 also known as Lymphocyte Activation Gene 3 (LAG-3). LAG-3 is a 70kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).

This 3DS223H antibody will recognize a formaldehyde-fixed epitope.
Description & Specifications

Specifications

CD223 (Lag-3)
Murine
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
3DS223H
Lymphocyte Activation Gene-3
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer