missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD223 (LAG-3), APC, clone: 3DS223H, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS17223942

 View more versions of this product

Catalog No. 501128842



Description

Description

LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).

Description: This 3DS223H monoclonal antibody recognizes human CD223 also known as Lymphocyte Activation Gene 3 (LAG-3). LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).This 3DS223H antibody will recognize a formaldehyde-fixed epitope.Applications Reported: This 3DS223H antibody has been reported for use in flow cytometric analysis.Applications Tested: This 3DS223H antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD223 (Lag-3)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P18627
CD223, LAG-3, Ly66
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
3DS223H
APC
LAG3
Liquid
LAG3
IgG1, kappa, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3902
Documents

For Research Use Only.