Please login to your online account to display your discounted pricing

CD218a (IL-18 Receptor alpha) Mouse anti-Human, FITC, Clone: H44, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $404.55

Specifications

Antigen CD218a (IL-18 Receptor alpha)
Clone H44
Host Species Mouse
Gene Alias IL-1Rrp
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010024

 
life technologies
LS11718342
100 tests Each for $404.55

501122157

 
life technologies
LS11718341
25 tests Each for $201.55
Description & Specifications

Specifications

Antigen CD218a (IL-18 Receptor alpha)
Clone H44
Host Species Mouse
Gene Alias IL-1Rrp
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The H44 antibody reacts with the interleukin-18 receptor alpha chain, also known as IL-1Rrp, an IL-1R family member. IL-18R alpha is expressed on Th1 cells, NK cell subset, neutrophils and IL-12-stimulated tonsillar B cells. By itself, IL-18R alpha binds IL-18 with low affinity and when complexed with IL-18R beta chain binds IL-18 with high affinity. The proinflammatory cytokine IL-18 and its receptor complex play an important role in the innate and autoimmune responses.