Please login to your online account to display your discounted pricing

CD218a (IL-18 Receptor alpha) Mouse anti-Human, APC, Clone: H44, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $419.05

Specifications

Antigen CD218a (IL-18 Receptor alpha)
Clone H44
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123187

 
life technologies
LS17718342
100 Tests Each for $419.05

501123186

 
life technologies
LS17718341
25 Tests Each for $216.05
Description & Specifications

Specifications

Antigen CD218a (IL-18 Receptor alpha)
Clone H44
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The H44 antibody reacts with the interleukin-18 receptor alpha chain, also known as IL-1Rrp, an IL-1R family member. IL-18R alpha is expressed on Th1 cells, NK cell subset, neutrophils and IL-12-stimulated tonsillar B cells. By itself, IL-18R alpha binds IL-18 with low affinity and when complexed with IL-18R beta chain binds IL-18 with high affinity. The proinflammatory cytokine IL-18 and its receptor complex play an important role in the innate and autoimmune responses.