missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD217 (IL-17 Receptor A) Mouse anti-Human, PE, Clone: J10MBS, eBioscience™

Mouse Monoclonal Antibody

$183.48 - $394.68

Specifications

Antigen CD217 (IL-17 Receptor A)
Clone J10MBS
Host Species Murine
Gene Alias Interleukin-17 receptor alpha
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011208 Life Technologies
LS12751741
25 Tests Each for $183.48
5011209 Life Technologies
LS12751742
100 Tests Each for $394.68
Specifications

Description

The J10MBS monoclonal antibody reacts with human interleukin 17 receptor A (IL-17RA, IL-17R). IL-17RA is a type I membrane protein of approximately 96kDa (866 amino acids) and is involved in inflammatory responses to IL-17A, IL-17F and IL-17AF heterodimer. It forms heterodimeric complexes with several other members of its receptor family, including IL-17RC for IL-17A and IL-17F signaling and IL-17RC for IL-17E (IL-25) signaling. IL-17RA is ubiquitously expressed, but in leukocytes its expression is elevated on granulocytes and monocytes.
Description & Specifications

Specifications

CD217 (IL-17 Receptor A)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
J10MBS
Interleukin-17 receptor alpha
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test