missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD217 (IL-17 Receptor A) Mouse anti-Human, Clone: 424LTS, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$196.68 - $460.68

Specifications

Antigen CD217 (IL-17 Receptor A)
Clone 424LTS
Host Species Murine
Gene Alias IL-17R, IL-17RA
Applications Functional Assay
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014764 Life Technologies
LS16791782
100μg Each for $196.68
5014765 Life Technologies
LS16791785
500μg Each for $460.68
Specifications

Description

The 424LTS monoclonal antibody reacts with and blocks IL-17A binding to human interleukin 17 receptor A (IL-17RA, IL-17R). IL-17RA is a type I membrane protein of approximately 96kDa (866 amino acids) and is involved in inflammatory responses to IL-17A, IL-17F and IL-17AF heterodimer. It forms heterodimeric complexes with several other members of its receptor family, including IL-17RC for IL-17A and IL-17F signaling and IL-17RC for IL-17E (IL-25) signaling. IL-17RA is ubiquitously expressed, but in leukocytes its expression is elevated on granulocytes and monocytes.
Description & Specifications

Specifications

CD217 (IL-17 Receptor A)
Murine
Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
424LTS
IL-17R, IL-17RA
RUO
Purified
Primary
aqueous buffer, no sodium azide