missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD217 (IL-17 Receptor A) Mouse anti-Human, APC, Clone: 424LTS, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $394.68

Specifications

Antigen CD217 (IL-17 Receptor A)
Clone 424LTS
Host Species Murine
Gene Alias IL-17RA
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015330 Life Technologies
LS17791741
25 Tests Each for $157.08
5015331 Life Technologies
LS17791742
100 Tests Each for $394.68
Specifications

Description

The 424LTS monoclonal antibody reacts with and blocks IL-17A binding to human interleukin 17 receptor A (IL-17RA, IL-17R). IL-17RA is a type I membrane protein of approximately 96kDa (866 amino acids) and is involved in inflammatory responses to IL-17A, IL-17F and IL-17AF heterodimer. It forms heterodimeric complexes with several other members of its receptor family, including IL-17RC for IL-17A and IL-17F signaling and IL-17RC for IL-17E (IL-25) signaling. IL-17RA is ubiquitously expressed, but in leukocytes its expression is elevated on granulocytes and monocytes.
Description & Specifications

Specifications

CD217 (IL-17 Receptor A)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
424LTS
IL-17RA
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test