Please login to your online account to display your discounted pricing

CD215 (IL-15 Receptor alpha) Rat anti-Mouse, PerCP-eFluor 710, Clone: DNT15Ra, eBioscience™

Rat Monoclonal Antibody

$187.05 - $462.55

Specifications

Antigen CD215 (IL-15 Receptor alpha)
Clone DNT15Ra
Host Species Rat
Gene Alias Interleukin-15 Receptor alpha, CD215
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123898

 
life technologies
LS46714980
25μg Each for $187.05

501123899

 
life technologies
LS46714982
100μg Each for $462.55
Description & Specifications

Specifications

Antigen CD215 (IL-15 Receptor alpha)
Clone DNT15Ra
Host Species Rat
Gene Alias Interleukin-15 Receptor alpha, CD215
Species Reactivity Mouse
Applications Flow Cytometry
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This DNT15Ra antibody reacts with the alpha subunit of the Mouse IL-15 Receptor. The IL-15 Receptor alpha subunit is unique to IL-15, while the other two subunits of the receptor, beta and gamma, are shared with IL-2. IL-15R signaling is mediated by Jak/STAT pathways, and results in the activation of neutrophils, dendritic cells, and macrophages. IL-15 has also been found to play a critical role in the development and survival of NK cells and CD8+ T cells.

Although high levels of mouse IL-15 mRNA have been observed in many cell types, free IL-15 is rarely detectable in biological samples. Recent research suggests that IL-15 is retained inside the cell and only secreted in complex with IL-15 Receptor alpha. This chaperoning is necessary from the ER to the Golgi, through secretion. The cytokine/receptor complex may stay bound to the cell surface, where it can be trans-presented to cells expressing the beta-gamma subunits of the receptor, or can be secreted in a soluble form lacking the transmembrane domain of IL-15 Receptor alpha. IL-15 and IL-15 Receptor alpha are expressed by dendritic cells and macrophages in response to inflammatory stimuli.

Our testing suggests that there may be strain differences in expression and staining of the Mouse IL-15 Receptor alpha. This DNT15Ra antibody has been tested on cells from C57Bl/6 mice. Staining has also been observed with this antibody after pre-incubation of NK cells with exogenous Mouse IL-15/IL-15R Complex, suggesting that DNT15Ra will recognize both Mouse IL-15 Receptor alpha and the Mouse IL-15/IL-15R Complex.