missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD21 Mouse anti-Human, FITC, Clone: HB5, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $317.55

Specifications

Antigen CD21
Clone HB5
Host Species Murine
Gene Alias CR2, Complement Component 3d Receptor (C3DR), EBV Receptor
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122047 Life Technologies
LS11021941
25 Tests Each for $129.05
501122048 Life Technologies
LS11021942
100 Tests Each for $317.55
Specifications

Description

The HB5 monoclonal antibody reacts with human CD21, an approximately 145kDa type I transmembrane protein. CD21 is expressed by mature B cells, folliculate dendritic cells and a subset of epithelial cells and is a receptor for complement component C3d and Epstein-Barr virus (EBV). CD21, in association with CD19 and CD81, participates in the multimolecular complex with BCR and is involved in B cell activation.
Description & Specifications

Specifications

CD21
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HB5
CR2, Complement Component 3d Receptor (C3DR), EBV Receptor
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test