Please login to your online account to display your discounted pricing

CD21 Mouse anti-Human, APC, Clone: HB5, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $419.05

Specifications

Antigen CD21
Clone HB5
Host Species Mouse
Gene Alias CR2, Complement Component 3d Receptor (C3DR), EBV Receptor
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014916

 
life technologies
LS17021941
25 tests Each for $201.55

5014917

 
life technologies
LS17021942
100 tests Each for $419.05
Description & Specifications

Specifications

Antigen CD21
Clone HB5
Host Species Mouse
Gene Alias CR2, Complement Component 3d Receptor (C3DR), EBV Receptor
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

The HB5 monoclonal antibody reacts with human CD21, an approximately 145kDa type I transmembrane protein. CD21 is expressed by mature B cells, folliculate dendritic cells and a subset of epithelial cells and is a receptor for complement component C3d and Epstein-Barr virus (EBV). CD21, in association with CD19 and CD81, participates in the multimolecular complex with BCR and is involved in B cell activation.