missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD21/CD35 Rat anti-Mouse, APC-eFluor 780, Clone: eBio8D9, eBioscience™

Rat Monoclonal Antibody

$157.08 - $407.88

Specifications

Antigen CD21/CD35
Clone eBio8D9
Host Species Rat
Gene Alias CR2/CR1, C3DR
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124012 Life Technologies
LS47021180
25μg Each for $157.08
501124013 Life Technologies
LS47021182
100μg Each for $407.88
Specifications

Description

The monoclonal antibody eBio8D9 reacts with an eptiope shared by mouse CD21 (CR2) and CD35 (CR1). CD21 and CD35 are alternatively spliced transcripts from the Cr2 gene, which produce cell-surface proteins of 145 and 190 kDa, respectively. CD21 and CD35 are expressed by mature B cells, but not on thymocytes, peripheral T cells, erythrocytes or platelets. Furthermore, there is some evidence which demonstrates their expression on macrophages. CD21 is a receptor for the complement component C3d and Epstein-Barr virus (EBV). In association with CD19 and CD81, CD21 also participates in B-cell activation through the B cell receptor. Cr2-deficient mice display impaired inflammatory and humoral immune responses in vivo.

The anti-mouse CD21/35 monoclonal antibody clones eBio4E3 and eBio8D9 do not cross-block each other, suggesting that they bind to different epitopes.
Description & Specifications

Specifications

CD21/CD35
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio8D9
CR2/CR1, C3DR
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL