Please login to your online account to display your discounted pricing

CD209 (DC-SIGN) Rat anti-Mouse, PE, Clone: 5H10, eBioscience™

Rat Monoclonal Antibody

$172.55 - $448.05

Specifications

Antigen CD209 (DC-SIGN)
Clone 5H10
Host Species Rat
Gene Alias DCSIGN, CIRE
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010722

 
affymetrix
12209180
25μg Each for $172.55

5010723

 
affymetrix
12209182
100μg Each for $448.05
Description & Specifications

Specifications

Antigen CD209 (DC-SIGN)
Clone 5H10
Host Species Rat
Gene Alias DCSIGN, CIRE
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 5H10 antibody was generated by immunization with a peptide derived from the extracellular region of mouse CIRE/DC-SIGN (CD209). CIRE/DC-SIGN was identified by its expression on CD8α- dendritic cells and plasmacytoid predendritic cells, and is the closest homologue of human DC-SIGN. Human DC-SIGN was originally identified in human placenta for its ability to bind the HIV envelope protein gp120 in a CD4-independent manner. CIRE/DC-SIGN is a 33kDa type II transmembrane C-type lectin protein. It contains a C-terminal, extracellular, Carbohydrate Recognition Domain (CRD) that is predicted to bind mannose and other carbohydrates in a Ca2+ dependent manner. It has been postulated that CIRE/DC-SIGN may play a role in T-dendritic cell interactions through binding with members of the ICAM family. CIRE/DC-SIGN is differentially expressed by sub-populations of dendritic cells and preliminary data suggest that its expression varies depending on the activation state of the host. CIRE/DC-SIGN is down-regulated in spleen-derived dendritic cell cultures supplemented with GM-CSF. While human DC-SIGN is predominantly expressed in dendritic cells, CIRE/DC-SIGN mRNA has also been detected in B cells. The 5H10 monoclonal antibody does not cross-react with the closely related SIGNR1, SIGNR2, SIGNR3 or SIGNR4.