missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD209 (DC-SIGN) Rat anti-Human, PE-Cy7, Clone: eB-h209, eBioscience™

Rat Monoclonal Antibody

$170.28 - $394.68

Specifications

Antigen CD209 (DC-SIGN)
Clone eB-h209
Host Species Rat
Gene Alias Clec4L
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123358 Life Technologies
LS25209942
100 Tests Each for $394.68
50245743 Life Technologies
LS25209941
25 Tests Each for $170.28
Specifications

Description

The eB-h209 monoclonal antibody reacts with human CD209, also known as DC-SIGN, a 44kDa type II transmembrane protein. DC-SIGN contains a C-type lectin binding domain and binds ICAM-3, ICAM-2, and HIV virus. Human dendritic cells preferentially express DC-SIGN. It has been postulated that DC-SIGN serves as a receptor for capture, trafficking, and transmission of HIV to T cells and supports primary immune response. eB-h209 was developed against a C-terminal peptide of human DC-SIGN.
Description & Specifications

Specifications

CD209 (DC-SIGN)
Rat
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.06μg)/test
eB-h209
Clec4L
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer