Please login to your online account to display your discounted pricing

CD209 (DC-SIGN) Rat anti-Human, Clone: eB-h209, eBioscience™

Rat Monoclonal Antibody

$201.55 - $375.55

Specifications

Antigen CD209 (DC-SIGN)
Clone eB-h209
Host Species Rat
Gene Alias Clec4L
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122663

 
life technologies
LS14209982
100μg Each for $375.55

5012988

 
life technologies
LS14209980
25μg Each for $201.55
Description & Specifications

Specifications

Antigen CD209 (DC-SIGN)
Clone eB-h209
Host Species Rat
Gene Alias Clec4L
Species Reactivity Human
Applications Flow Cytometry
Applications Immunoprecipitation
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The eB-h209 monoclonal antibody reacts with human CD209, also known as DC-SIGN, a 44kDa type II transmembrane protein. DC-SIGN contains a C-type lectin binding domain and binds ICAM-3, ICAM-2, and HIV virus. Human dendritic cells preferentially express DC-SIGN. It has been postulated that DC-SIGN serves as a receptor for capture, trafficking, and transmission of HIV to T cells and supports primary immune response. eB-h209 was developed against a C-terminal peptide of human DC-SIGN.