missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD209 (DC-SIGN) Rat anti-Human, Biotin, Clone: eB-h209, eBioscience™

Rat Monoclonal Antibody

$183.48 - $368.28

Specifications

Antigen CD209 (DC-SIGN)
Clone eB-h209
Host Species Rat
Gene Alias Clec4L
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122531 Life Technologies
LS13209982
100μg Each for $368.28
5011826 Life Technologies
LS13209980
25μg Each for $183.48
Specifications

Description

The eB-h209 monoclonal antibody reacts with human CD209, also known as DC-SIGN, a 44kDa type II transmembrane protein. DC-SIGN contains a C-type lectin binding domain and binds ICAM-3, ICAM-2, and HIV virus. Human dendritic cells preferentially express DC-SIGN. It has been postulated that DC-SIGN serves as a receptor for capture, trafficking, and transmission of HIV to T cells and supports primary immune response. eB-h209 was developed against a C-terminal peptide of human DC-SIGN.
Description & Specifications

Specifications

CD209 (DC-SIGN)
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
eB-h209
Clec4L
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer