Please login to your online account to display your discounted pricing

CD209 (DC-SIGN), CD209a Mouse anti-Mouse, eFluor 660, Clone: MMD3, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen CD209 (DC-SIGN), CD209a
Clone MMD3
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124324

 
life technologies
LS50209480
25μg Each for $172.55

501124325

 
life technologies
LS50209482
100μg Each for $433.55
Description & Specifications

Specifications

Antigen CD209 (DC-SIGN), CD209a
Clone MMD3
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This MMD3 monoclonal antibody recognizes mouse CD209a, which is also known as DC-SIGN. CD209a is a type II transmembrane C-type lectin expressed on a subset of dendritic cells, including some CD4+, CD8- and plasmacytoid pre-dendritic cells. Studies indicate that CD209a expression can vary according to the activation state of the host. Moreover, CD209a is down-regulated in spleen-derived dendritic cell cultures supplemented with GM-CSF. CD209a is involved in mediating the innate immune response by binding microbial carbohydrates.

Cross-blocking studies suggest that MMD3 recognizes a different a different epitope from LWC06 and 5H10.