missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD207 (Langerin) Rat anti-Mouse, Clone: eBioRMUL.2, eBioscience™

Rat Monoclonal Antibody

$117.48 - $341.88

Specifications

Antigen CD207 (Langerin)
Clone eBioRMUL.2
Host Species Rat
Gene Alias Clec4K
Applications Flow Cytometry (Intracellular Staining),Immunohistochemistry,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012978 Life Technologies
LS14207380
25μg Each for $117.48
5012979 Life Technologies
LS14207382
100μg Each for $341.88
Specifications

Description

RMUL.2 is a monoclonal antibody that reacts with the predicted extracellular domain of mouse CD207, also known as Langerin. Mouse CD207 is expressed in a distinct subset of dendritic cells called Langerhans cells (LC). Mouse CD207 is a 48 kDa, C-type lectin and putative transmembrane protein that likely plays a role in antigen recognition and uptake. LC are located in the epidermis and upon activation reduce expression of CD207 and begin migrating through the dermis towards lymphatic vessels. Expression of mouse CD207 in LC has been correlated to the presence of Birbeck Granules. Furthermore, the formation of Birbeck Granules has been observed upon transfection of CD207 cDNA into fibroblast cell lines.

Western Blotting with RMUL.2 reveals a 48kDa band in lysate from mouse ear epidermis as well as in lysates from cells transfected with mouse CD207 cDNA. Cross reactivity of this antibody with other proteins has not been determined. Preliminary data from mouse CD207 cDNA transfected cells suggests that RMUL.2 can be used for intracellular staining followed by flow cytometric analysis. We have observed in our transfectants the majority of protein is expressed intracellularly with a small percentage on the cell surface. In addition, RMUL.2 has also been tested by immunohistology on mouse ear epidermal sheets and shows specific staining.
Description & Specifications

Specifications

CD207 (Langerin)
Rat
Flow Cytometry (Intracellular Staining),Immunohistochemistry,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
eBioRMUL.2
Clec4K
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer