missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD207 (Langerin) Rat anti-Mouse, Clone: eBioL31, eBioscience™

Rat Monoclonal Antibody

$117.48 - $328.68

Specifications

Antigen CD207 (Langerin)
Clone eBioL31
Host Species Rat
Gene Alias Clec4K
Applications Flow Cytometry,Flow Cytometry (Intracellular Staining),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122662 Life Technologies
LS14207582
100μg Each for $328.68
5012980 Life Technologies
LS14207580
25μg Each for $117.48
Specifications

Description

The eBioL31 monoclonal antibody reacts with mouse CD207, also known as Langerin, which is expressed in a distinct subset of dendritic cells called Langerhans cells (LC). Mouse CD207 is a 48kDa C-type lectin transmembrane protein that likely plays a role in antigen recognition and uptake. LC are located in the epidermis and upon activation, reduce CD207 expression and begin migrating through the dermis towards lymphatic vessels. Expression of mouse CD207 in LC has been correlated to the presence of Birbeck Granules. Furthermore, the formation of Birbeck Granules has been observed upon transfection of CD207 cDNA into fibroblast cell lines.

Western Blotting with eBioL31 reveals a 48kDa band in lysate from mouse ear epidermis, as well as in lysates from cells transfected with mouse CD207 cDNA. Preliminary data from mouse CD207 cDNA transfected cells suggests that eBioL31 can be used for intracellular flow cytometric analysis in combination with the Foxp3/Transcription Factor Staining Bufer Set (cat. 00-5523). The epitope recognized by eBioL31 is in the extracellular domain within the CRD (carbohydrate recognition domain) and therefore can be used to detect both surface and intracellular langerin. Differences in expression has been observed between mouse strains.
Description & Specifications

Specifications

CD207 (Langerin)
Rat
Flow Cytometry,Flow Cytometry (Intracellular Staining),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
eBioL31
Clec4K
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer