missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD205 Rat anti-Mouse, PerCP-eFluor 710, Clone: 205yekta, eBioscience™

Rat Monoclonal Antibody

$170.28 - $170.28

Specifications

Antigen CD205
Clone 205yekta
Host Species Rat
Gene Alias DEC205, Ly75
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016027 Life Technologies
LS46205180
25μg Each for $170.28
Specifications

Description

The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells.

CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells.
Description & Specifications

Specifications

CD205
Rat
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
205yekta
DEC205, Ly75
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer