missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD205 Rat anti-Mouse, PE-eFluor 610, Clone: 205yekta, eBioscience™

Rat Monoclonal Antibody

$216.05 - $535.05

Specifications

Antigen CD205
Clone 205yekta
Host Species Rat
Gene Alias DEC-205, Ly75
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Qty  
501124795 Life Technologies
LS61205180
25μg Each for $216.05
501124796 Life Technologies
LS61205182
100μg Each for $535.05
Specifications

Description

The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells.

CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells.
Description & Specifications

Specifications

CD205
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
205yekta
DEC-205, Ly75
Flow Cytometry
PE-eFluor 610
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL