missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD205 Rat anti-Mouse, PE-eFluor® 610, Clone: 205yekta, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS61205182

Catalog No. 501124796



Description

Description

Description: The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of

Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD205 (LY25, Lymphocyte Antigen 75) is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 acts as an endocytic receptor that directs captured antigens to the extracellular space towards a specialized antigen processing region. Further, CD205 also prompts the reduced proliferation of B lymphocytes. Diseases associated with CD205 dysfunction include pneumonic plague and lipid pneumonia.
Specifications

Specifications

CD205
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q60767
RP23-10L17.4, CD205, DEC-205, ly-75, CLEC13B, GP200-MR6, DCL-1 fusion, LY75-CD302 fusion protein
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
205yekta
PE-eFluor® 610
Ly75
Liquid
Ly75
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
17076
Documents

For Research Use Only.