Learn More
CD205 Monoclonal Antibody (205yekta), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$221.00 - $493.00
Specifications
Antigen | CD205 |
---|---|
Clone | 205yekta |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Description: The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells. Applications Reported: This 205yekta antibody has been reported for use in flow cytometric analysis. Applications Tested: This 205yekta antibody has been pre-titrated and tested by flow cytometric analysis of in vitro-cultured bone marrow-derived dendritic cells or mouse Granulocytes. This can be used at 5 μL (0.007 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.
CD205 Monoclonal antibody specifically detects CD205 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD205 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q60767 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
205yekta | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
17076 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.