Please login to your online account to display your discounted pricing

CD205 Rat anti-Mouse, PE, Clone: 205yekta, eBioscience™

Rat Monoclonal Antibody

$172.55 - $361.05

Specifications

Antigen CD205
Clone 205yekta
Host Species Rat
Gene Alias DEC-205, Ly75
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010716

 
affymetrix
12205180
25μg Each for $172.55

5010717

 
affymetrix
12205182
100μg Each for $361.05
Description & Specifications

Specifications

Antigen CD205
Clone 205yekta
Host Species Rat
Gene Alias DEC-205, Ly75
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells.

CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells.