missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD205 Rat anti-Mouse, Biotin, Clone: 205yekta, eBioscience™

Rat Monoclonal Antibody

$183.48 - $407.88

Specifications

Antigen CD205
Clone 205yekta
Host Species Rat
Gene Alias DEC-205, Ly75
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122530 Life Technologies
LS13205182
100μg Each for $407.88
5011816 Life Technologies
LS13205180
25μg Each for $183.48
Specifications

Description

The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells.

CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells.
Description & Specifications

Specifications

CD205
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
205yekta
DEC-205, Ly75
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer