Please login to your online account to display your discounted pricing

CD203c Mouse anti-Human, APC, Clone: NP4D6, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $442.25

Specifications

Antigen CD203c
Clone NP4D6
Host Species Murine
Gene Alias E-NPP3, PDNP3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123101

 
life technologies
LS17203941
25 Tests Each for $187.05

501123102

 
life technologies
LS17203942
100 Tests Each for $442.25
Description & Specifications

Specifications

Antigen CD203c
Clone NP4D6
Host Species Murine
Gene Alias E-NPP3, PDNP3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This NP4D6 monoclonal antibody reacts with human CD203c, which is also known as ectonucleotide pyrophosphatase/phosphodiesterase 3 (E-NPP3). ENPP family members catalyze the hydrolysis of pyrophosphate and phosphodiester bonds to nucleoside 5-monophosphates. A type II transmembrane protein, CD203c is expressed on basophils and mast cells. CD203c expression is upregulated on basophils activated by allergen or IgE crosslinking. Therefore, CD203c is used as an indicator of basophil activation in various IgE-mediated allergic responses.